Learn More
Medchemexpress LLC SAG | 912545-86-9 | 99.60% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY12848100MG
Encompass_Preferred
SAG is a potent Smoothened (Smo) receptor agonist that activates the Hedgehog signaling pathway. It can counteract the inhibition of Smo by Cyclopamine and is intended for research use only.
  • Potent Smoothened (Smo) receptor agonist (EC50=3 nM; Kd=59 nM)
  • Activates the Hedgehog signaling pathway
  • Counteracts inhibition of Smo
  • For research use only
Catalog No. 50-003-42013