Learn More
Medchemexpress LLC SAG-d3 | 912545-86-9 | 99.9% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY12848S1MG
Encompass_Preferred
SAG-d3 is deuterium labeled SAG. SAG is a potent Smoothened (Smo) receptor agonist with an EC50 of 3 nM and a Kd of 59 nM.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-39985