Learn More
Medchemexpress LLC SAG dihydrochloride | 2702366-44-5 | 98.2% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY12848C100MG
Encompass_Preferred
SAG dihydrochloride is a potent Smoothened (Smo) receptor agonist with an EC50 of 3 nM and a Kd of 59 nM. It activates the Hedgehog signaling pathway and counteracts Cyclopamine inhibition of Smo, making it suitable for various research applications.
  • Potent Smoothened (Smo) receptor agonist
  • Activates the Hedgehog signaling pathway
  • Counteracts Cyclopamine inhibition of Smo
  • Induces firefly luciferase expression in Shh-LIGHT2 cells
  • Competes for binding of BODIPY-cyclopamine to Smo-expressing Cos-1 cells
  • Increases SMO mRNA and protein expression in MDAMB231 cells
Catalog No. 50-003-41842