Learn More
Cayman Chemical SAHA, 149647-78-9, 100 mg
SDP
Supplier:  Cayman Chemical 10009929100
Encompass_Preferred
Molecular Formula C14H20N2O3, Purity ≥98%, Formula Weight 264.3, Synonyms: Suberoylanilide Hydroxamic Acid, Vorinostat; An inhibitor of class I, II, and IV HDACs at 50-200 nM; arrests cell growth in a wide variety of transformed cells in culture at 2.5-5.0 M.
Catalog No. NC9626646