Learn More
Abcam SAHA (Vorinostat), HDAC inhibitor, 10MG
Supplier:  Abcam AB14448010MG
Encompass_Preferred
MW 264.32 Da, Purity >98%. Potent non-selective HDAC inhibitor (IC₅₀ values are 10 and 20 nM for HDAC1 and HDAC3, respectively). Induces apoptosis and shows antiproliferative effects against cancer cell lines (IC₅₀ = 3 - 8 μM). Antitumor effects in vivo. No effect on class III HDACs. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5680