Learn More
Medchemexpress LLC SAHM1 | 2050906-89-1 | 96.4% | 2196.58 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP220310MG
Encompass_Preferred
SAHM1 is a peptide mimetic of a dominant negative form of mastermind-like (MAML). It inhibits canonical Notch transcription complex formation and can be used for the research of allergic airway inflammation in mice.
  • Inhibits canonical Notch transcription complex formation.
  • Used for research of allergic airway inflammation in mice.
  • Has an IC50 for Notch.
Catalog No. 50-004-22183