Learn More
Cayman Chemical SaIkosaponIn F 5mg
SDP
Supplier:  Cayman Chemical 356265 MG
Encompass_Preferred
A triterpenoid saponin and an antagonist of TRPA1; inhibits polygodial- or AITC-induced calcium flux in HEK293 cells expressing human TRPA1 from 0.5-1.5 µM
Catalog No. 50-291-3663