Learn More
Pacer Global LLC Saint-Gobain - 082 C-Flex TPE Tubing, 0.063'' ID, 0.188'' OD, 0.0625'' Wall, 50 Foot Coil, 1/EA - 082-063-2
SDP
Supplier:  Pacer Global LLC MLS0820632
Encompass_Preferred
Saint-Gobain - 082 C-Flex TPE Tubing, 0.063" ID, 0.188" OD, 0.0625" Wall, 50 Foot Coil, 1/EA
Catalog No. 50-003-32300