Learn More
Pacer Global LLC Saint-Gobain - 374 C-Flex TPE Tubing, 0.375'' ID, 0.625'' OD, 0.125'' Wall, 150 Foot Coil, 1/EA - 374-375-4M
SDP
Supplier:  Pacer Global LLC MLS3743754M
Encompass_Preferred
Saint-Gobain - 374 C-Flex TPE Tubing, 0.375" ID, 0.625" OD, 0.125" Wall, 150 Foot Coil, 1/EA
Catalog No. 50-003-32294