Learn More
Pacer Global LLC Saint-Gobain - 374 C-Flex TPE Tubing, 0.5'' ID, 0.75'' OD, 0.125'' Wall, 250 Foot Coil, 1/EA - 374-500-4S
SDP
Supplier:  Pacer Global LLC MLS3745004S
Encompass_Preferred
Saint-Gobain - 374 C-Flex TPE Tubing, 0.5" ID, 0.75" OD, 0.125" Wall, 250 Foot Coil, 1/EA
Catalog No. 50-003-32472