Learn More
Accuflow Systems Saint-Gobain BarbLock Hand Tool, Steel, 625 Series, 1 ea
Supplier: Accuflow Systems BLT135625
The Saint-Gobain BarbLock® Hand Tool is designed for low-volume assembly of BarbLock® retainers, securing flexible tubing and hose onto barbed fittings. It is available in steel and PVDF versions.
- Designed for low-volume assembly of BarbLock® retainers
- Secures flexible tubing and hose onto barbed fittings
- Creates a leak-free connection with 360° radial crimp and compression
- Compatible with 0.625 in. tubing inner diameter