Learn More
Accuflow Systems Saint-Gobain BarbLock Retainer, 0.250 in. ID, Polypropylene, 1 ea
Supplier: Accuflow Systems BL135252
The BarbLock® retainer is designed for retaining flexible tubing and hose onto barbed fittings. It provides full 360° radial crimp behind the barb and 360° of compression directly over the barb's ridge, eliminating leak paths. Made from high-performance polypropylene and PVDF resins, it requires no adhesives or solvents, making it suitable for sensitive liquid and gas applications.
- Compatible with all flexible tubing materials.
- Provides full 360° radial crimp behind the barb.
- Provides 360° compression directly on the barb's ridge.
- Eliminates solvent bonding, ultraviolet bonding, and toxic adhesives.
- Manufactured from resins that meet the requirements of USP <88> Class VI, and/or USP <87>, and/or ISO 10993-5.
- Custom color and identification options available.
- Quick and easy to assemble.