Learn More
Accuflow Systems Saint-Gobain BarbLock Retainer, 0.375 x 0.562 in., PP, 1 ea
Supplier:  Accuflow Systems BL135374
Encompass_Preferred
BarbLock® retainers are designed to replace inconsistent, leak-prone cable ties and traditional clamps for securing flexible tubing and hose onto barbed fittings. They feature full 360° radial crimp and compression to prevent leaks. Constructed from high-performance polypropylene, these retainers meet USP <88> Class VI, USP <87>, and/or ISO 10993-5. They require no adhesives or solvents and offer quick, easy, leak-free assembly.
  • Compatible with all flexible tubing materials
  • Full 360° radial crimp behind the barb
  • Provides 360° compression directly on the barb's ridge
  • Eliminates solvent bonding, ultraviolet bonding and toxic adhesives
  • Manufactured from resins that meet USP <88> Class VI, USP <87>, and/or ISO 10993-5 requirements
  • Quick and easy to assemble
Catalog No. NC3936018