Learn More
Accuflow Systems Saint-Gobain BarbLock Retainer, 0.500 in. ID, Polypropylene, 1 ea
Supplier:  Accuflow Systems BL135502
Encompass_Preferred
BarbLock® Ultra-Secure Retainers are advanced polypropylene devices designed for securing flexible tubing onto barbed fittings. They feature a full 360° radial crimp and compression to prevent leaks in demanding fluid transfer applications, meeting USP Class VI and ISO 10993-5 requirements.
  • Replaces inconsistent cable ties and traditional clamps.
  • Compatible with all flexible tubing materials and Pure-Fit® SIB® fittings.
  • Provides 360° radial crimp behind the barb and full dynamic retention.
  • Provides 360° compression directly on the barb's ridge.
  • Eliminates solvent bonding, ultraviolet bonding, and toxic adhesives.
  • Quick and easy assembly.
Catalog No. NC3936003