Learn More
Accuflow Systems Saint-Gobain BarbLock Retainer, Polypropylene, 0.250 in. ID, 1 ea
Supplier: Accuflow Systems BL135251AL
BarbLock retainers provide an advanced solution for securing flexible tubing and hose onto barbed fittings, replacing cable ties and traditional clamps. Featuring a full 360° radial crimp and compression over the barb's ridge, they solve leak path problems in fluid transfer. Manufactured from high-performance polypropylene, they meet USP <88> Class VI, USP <87>, and/or ISO 10993-5 requirements.
- Compatible with all flexible tubing materials.
- Full 360° radial crimp behind the barb.
- Direct 360° compression on the barb's ridge.
- Eliminates solvent bonding and toxic adhesives.
- Resins meet USP <88> Class VI, USP <87>, and/or ISO 10993-5 standards.
- Ensures quick and easy assembly.
- AL series one-piece design integrates the sleeve and collet.
- Gamma stable.