Learn More
Accuflow Systems Saint Gobain BarbLock Retainers, 1/2 in. ID, 250/Pk.
Supplier:  Accuflow Systems BL135503
Encompass_Preferred
The BarbLock® retainer is designed to secure flexible tubing and hose to barbed fittings in critical fluid transfer applications, providing a 360° radial crimp and compression to prevent leaks. Made from high-performance, USP Class VI compliant polypropylene and PVDF resins, these retainers are gamma stable and compatible with various tubing and fittings.
  • Provides 360° radial crimp behind the barb and 360° compression over the barb's ridge
  • Made from high-performance polypropylene and PVDF resins
  • Meets USP Class VI standards
  • Compatible with all flexible tubing materials and SIB® fittings
  • Gamma stable for various sterilization methods
  • Quick and easy, tool-free assembly
  • Eliminates solvent bonding, ultraviolet bonding, and toxic adhesives
Catalog No. NC3936006