Learn More
Pacer Global LLC Saint Gobain PharMed BPT Tubing, 0.375" ID, 0.5" OD, 0.0625" Wall, 25 Foot Coil, 1/EA
SDP
Supplier:  Pacer Global LLC MLSAY242027
Encompass_Preferred
Saint Gobain PharMed BPT Tubing, 0.375" ID, 0.5" OD, 0.0625" Wall, 25 Foot Coil, 1/EA
Catalog No. 50-003-32350