Learn More
Pacer Global LLC Saint-Gobain - SaniTech Ultra-65 Tubing, 0.25'' ID, 0.375'' OD, 0.0625'' Wall, 50 Foot Coil, 1/EA - Ultra65-250-2F
SDP
Supplier:  Pacer Global LLC MLSULTRA652502F
Encompass_Preferred
Saint-Gobain - SaniTech Ultra-65 Tubing, 0.25" ID, 0.375" OD, 0.0625" Wall, 50 Foot Coil, 1/EA
Catalog No. 50-003-32394