Learn More
Pacer Global LLC Saint-Gobain - SaniTech Ultra-C Tubing, 0.5'' ID, 0.75'' OD, 0.125'' Wall, 50 Foot Coil, 1/EA - ULTRA-C-500-4F
SDP
Supplier:  Pacer Global LLC MLSULTRAC5004F
Encompass_Preferred
Saint-Gobain - SaniTech Ultra-C Tubing, 0.5" ID, 0.75" OD, 0.125" Wall, 50 Foot Coil, 1/EA
Catalog No. 50-003-32427