Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, 1 in ID, 1-3/8 in OD, Phthalate-free
Supplier:  Accuflow Systems AAA00064
Encompass_Preferred
Tygon B-44-4X tubing from Saint-Gobain is a food- and dairy-grade flexible tubing designed for sanitary filling, draining, transfer, and processing applications. It provides excellent clarity for visual inspection, a smooth nonporous bore for hygienic fluid handling, and formulations free of BPA and phthalates.
  • 1 in inside diameter (25.40 mm)
  • 1-3/8 in outside diameter (34.93 mm)
  • 3/16 in wall thickness (0.1875 in, 4.76 mm)
  • Phthalate-free formulation
  • Suitable for food, milk, and dairy transfer and processing
  • Complies with FDA food additive regulations, NSF-51, NSF-61, and 3-A
  • Working and burst pressures are test-derived; verify suitability under application conditions
Catalog No. NC3909048