Learn More
Medchemexpress LLC Sakuranetin | 2957-21-3 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN30061MG
Encompass_Preferred
Sakuranetin is a cherry flavonoid phytoalexin known for its strong antifungal, anti-inflammatory, and antioxidative properties. It has been shown to ameliorate LPS-induced acute lung injury.
  • Exhibits strong antifungal activity
  • Possesses anti-inflammatory and antioxidative activities
  • Ameliorates LPS-induced acute lung injury
  • Inhibits fungal mycelium growth
  • Inhibits influenza B/Lee/40 virus replication
  • Induces melanin synthesis and increases the expression of Tyr, TRP1, and TRP2 in B16 melanoma cells
  • Inhibits the phosphorylation of ERK and AKT in B16 melanoma cells
  • Reduces inflammation and respiratory system elastance in LPS-induced acute lung inflammation models
  • Attenuates airway hyperresponsiveness and inflammation in Ovalbumin-induced asthma models
Catalog No. 50-004-21750