Learn More
Medchemexpress LLC Salbutamol | 18559-94-9 | 99.53% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB1037100MG
Encompass_Preferred
Salbutamol (Albuterol) is an orally active short-acting β2-adrenergic receptor agonist. It promotes tumorigenesis in gastric cancer cells through the β2-AR/ERK/EMT pathway. It can relax bronchial smooth muscle and is used to study bronchospasm induced by asthma and chronic obstructive pulmonary disease.
  • Orally active short-acting β2-adrenergic receptor agonist
  • Relaxes bronchial smooth muscle
  • Used to study bronchospasm induced by asthma and chronic obstructive pulmonary disease
Catalog No. 50-004-14194