Learn More
Medchemexpress LLC Salbutamol | 18559-94-9 | 99.9% | 10 MG

Supplier: Medchemexpress LLC HYB1037R10MG
Salbutamol (standard) is an analytical reference standard of salbutamol (albuterol) provided for laboratory testing, method development, and quality control. It is supplied as a small, high-purity vial intended for assay calibration and analytical reference use.
- High purity suitable for analytical applications.
- Provided in a small pack size for convenient laboratory use.
- Intended for method development, quality control, and assay calibration.
- Molecular formula C13H21NO3 and molecular weight 239.31 g/mol.
- CAS number 18559-94-9 for unambiguous identification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.