Learn More
Cayman Chemical Salbutamol-d9 1mg
SDP
Supplier:  Cayman Chemical 252341 MG
Encompass_Preferred
An internal standard for the quantification of salbutamol by GC- or LC-MS
Catalog No. 50-293-2578