Learn More
Cayman Chemical Salbutamol-d9 500ug
SDP
Supplier:  Cayman Chemical 25234500 UG
Encompass_Preferred
An internal standard for the quantification of salbutamol by GC- or LC-MS
Catalog No. 50-293-2538