Learn More
Medchemexpress LLC Salbutamol-d9 acetate (Albuterol-d9 acetate) | 1781417-68-2 | 99.5% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB1037S31MG
Encompass_Preferred
Salbutamol-d9 (Albuterol-d9) acetate is the deuterium-labeled form of Salbutamol, an orally active, short-acting β2-adrenergic receptor agonist. It promotes tumorigenesis in gastric cancer cells via the β2-AR/ERK/EMT pathway and can relax bronchial smooth muscle, used in studies of bronchospasm induced by asthma and chronic obstructive pulmonary disease. It is for research use only. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes into drug molecules for quantitation during drug development
  • May affect pharmacokinetic and metabolic profiles of drugs through deuteration
Catalog No. 50-004-13853