Learn More
Medchemexpress LLC Salbutamol hemisulfate | 51022-70-9 | 98.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB043610MM/1ML
Encompass_Preferred
Salbutamol hemisulfate (Albuterol hemisulfate) is an orally active short-acting β2-adrenergic receptor agonist. It promotes tumorigenesis in gastric cancer cells through the β2-AR/ERK/EMT pathway. Salbutamol hemisulfate can relax bronchial smooth muscle and is used to study bronchospasm induced by asthma and chronic obstructive pulmonary disease.
  • Orally active short-acting β2-adrenergic receptor agonist
  • Promotes tumorigenesis in gastric cancer cells through the β2-AR/ERK/EMT pathway
  • Relaxes bronchial smooth muscle
  • Used to study bronchospasm induced by asthma and chronic obstructive pulmonary disease
Catalog No. 50-004-08945