Learn More
Medchemexpress LLC Salbutamol (hemisulfate) (standard) | 51022-70-9 | 99.8% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0436R
Encompass_Preferred
Salbutamol hemisulfate is an analytical standard used in research and analytical applications. It acts as an orally active short-acting β2-adrenergic receptor agonist, relaxing bronchial smooth muscle. This compound is suitable for studying bronchospasm induced by asthma and chronic obstructive pulmonary disease, and it promotes tumorigenesis in gastric cancer cells through the β2-AR/ERK/EMT pathway.
  • Intended for research and analytical applications
  • Orally active short-acting β2-adrenergic receptor agonist
  • Promotes tumorigenesis in gastric cancer cells through the β2-AR/ERK/EMT pathway
  • Relaxes bronchial smooth muscle
  • Used to study bronchospasm induced by asthma and chronic obstructive pulmonary disease
  • Reference standard supplied assay
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-50593