Learn More
Medchemexpress LLC Salbutamol (Standard) | 18559-94-9 | 99.9% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB1037R100MG
Encompass_Preferred
Salbutamol (Standard) is the analytical standard of Salbutamol, intended for research and analytical applications. Salbutamol (Albuterol) is a short-acting beta-2 adrenergic receptor agonist with oral activity. It promotes tumorigenesis of gastric cancer cells through the β2-AR/ERK/EMT pathway and is used to study bronchospasms caused by asthma and chronic obstructive pulmonary disease (COPD).
  • Analytical standard, suitable for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Appearance: Solid, white to off-white.
  • Shipping: Room temperature in continental US; may vary elsewhere.
Catalog No. 50-004-13979