Learn More
Medchemexpress LLC Salcaprozate sodium | 1G
SDP
Supplier:  Medchemexpress LLC HY1142991G
Encompass_Preferred
Salcaprozate sodium (SNAC) an oral absorption promoter and has the potential as a delivery agent for oral forms of heparin and insulin Salcaprozate sodium could increase passive transcellular permeation across small intestinal epithelia based on increased lipophilicity arising from non-covalent macromolecule complexation[1][2]
Catalog No. 50-002-28089