Learn More
Medchemexpress LLC Salcaprozate sodium 250mg | 203787-91-1 | 250 MG
SDP
Supplier:  Medchemexpress LLC HY114299250MG
Encompass_Preferred
Salcaprozate (sodium) is the sodium salt form of salcaprozate, an oral absorption promoter used in formulation research to enhance gastrointestinal uptake of macromolecules. It increases passive transcellular permeation and is commonly used in studies of peptide and heparin delivery.
  • Enhances oral absorption of macromolecules by increasing intestinal permeability.
  • Used as an excipient or permeation enhancer in formulation studies.
  • Sodium salt, water-soluble and suitable for in vitro and in vivo research models.
  • Characterized by CAS 203787-91-1 and formula C15H20NNaO4.
Catalog No. 50-003-08127