Learn More
Medchemexpress LLC Salcomine (N,N′-Bis(salicylidene)ethylenediaminocobalt(II)) | 14167-18-1 | 99.1% | 325.23 | 50 G

Supplier: Medchemexpress LLC HYW01379450G
Salcomine is a carrier of O2 and oxidation catalysts, and has anti-influenza virus activity.
- Carrier of O2 and oxidation catalysts
- Anti-influenza virus activity
- Appearance: solid, brown to reddish brown
- Solubility: in DMSO at 2 mg/mL (6.15 mM) with ultrasonic, warming, and heating to 60°C
- Shipping: room temperature in continental US; may vary elsewhere
- Storage: 4°C, protect from light, stored under nitrogen. In solvent: -80°C for 6 months; -20°C for 1 month (protect from light, stored under nitrogen)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.