Learn More
Medchemexpress LLC Salicyl-AMS | 863238-55-5 | 99.6% | 466.43 | 100 MG

Supplier: Medchemexpress LLC HY10894110MM/1ML
Salicyl-AMS is a mycobactin biosynthesis inhibitor, effective in inhibiting *M. tuberculosis* growth in vitro under iron-limited conditions. This compound is intended for research applications only.
- Inhibits mycobactin biosynthesis
- Inhibits *M. tuberculosis* growth
- High purity: 99.59%
- Soluble in DMSO for in vitro studies
- Stable under specific storage conditions
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.