Learn More
Medchemexpress LLC Salicylamide | 65-45-2 | 99.9% | 100 G
SDP
Supplier:  Medchemexpress LLC HYB0811100G
Encompass_Preferred
Salicylamide is an inhibitor of microsomal UDP-glucuronosyltransferase. It also acts as an analgesic and anti-pyretic agent. Treatment with salicylamides leads to bacterial growth inhibition, which correlates with the level of inhibition of sulfate reduction.
  • Inhibits microsomal UDP-glucuronosyltransferase.
  • Acts as an analgesic and anti-pyretic agent.
  • Inhibits bacterial growth by reducing sulfate.
  • Decreases radiosulfate levels in maternal serum and placenta.
  • Impairs incorporation into fetal skeletal glycosaminoglycans.
  • Decreases calcium content in fetal limb bones.
  • Reduces radiosulfate uptake by maternal serum, liver, fetus, and placenta.
  • Soluble in DMSO (100 mg/mL) and H2O (0.1 mg/mL).
  • Solid appearance, off-white to light brown color.
Catalog No. 50-004-09143