Learn More
Medchemexpress LLC Salicylamide | 65-45-2 | 99.9% | 50 G
SDP
Supplier:  Medchemexpress LLC HYB081150G
Encompass_Preferred
Salicylamide, also known as 2-Hydroxybenzamide, is a chemical compound primarily recognized as an inhibitor of microsomal UDP-glucuronyltransferase. It also exhibits analgesic and anti-pyretic properties, making it a valuable compound for research in areas related to pain and fever management. This high-purity compound is intended for research use only.
  • Inhibits microsomal UDP-glucuronyltransferase
  • Acts as an analgesic agent
  • Functions as an anti-pyretic agent
  • High purity for reliable research
  • Suitable for various research applications
Catalog No. 50-004-09295