Learn More
Medchemexpress LLC Salicylamide | 65-45-2 | 99.9% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0811500MG
Encompass_Preferred
Salicylamide is an inhibitor of microsomal UDP-glucuronyltransferase and acts as an analgesic and anti-pyretic agent. It leads to bacterial growth inhibition and decreases levels of radiosulfate in maternal serum and placenta, impairing the incorporation of radiosulfate into fetal skeletal GAGs.
  • Inhibitor of microsomal UDP-glucuronosyltransferase.
  • Analgesic and anti-pyretic agent.
  • Leads to bacterial growth inhibition.
  • Decreases levels of radiosulfate in maternal serum and placenta.
  • For research use only; not sold to patients.
Catalog No. 50-004-07520