Learn More
Cayman Chemical SalIcylcurcumIn 10mg
SDP
Supplier:  Cayman Chemical 1187810 MG
Encompass_Preferred
A synthetic curcuminoid with antioxidant and anticarcinogenic properties; increases the activity of quinone reductase in mouse hepatoma cells in a dose-dependent fashion, doubling activity at 0.3 μM salicylcurcumin
Catalog No. 50-295-1653