Learn More
Medchemexpress LLC Salicylhydroxamic acid | 89-73-6 | 99.9% | 153.14 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1872110MM/1ML
Encompass_Preferred
Salicylhydroxamic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • Exhibits in vitro inhibitory activity against 5-lipoxygenase in rat basophilic leukemia cells (RBL-1).
  • Has an IC50 of 30 μM for 5-lipoxygenase inhibition.
Catalog No. 50-003-86116