Learn More
TARGETMOL CHEMICALS INC SALIDROSIDE 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2717
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Salidroside (Rhodioloside) is a bioactive phenolic glycoside compound isolated from Rhodiola crenulata. It is a prolyl endopeptidase Inhibitor. purity: 99%
Catalog No. 50-258-0640