Learn More
Selleck Chemical LLC Salidroside 10mg 10338-51-9 Rhodioloside
SDP
Supplier:  Selleck Chemical LLC S239610MG
Encompass_Preferred
Salidroside (Rhodioloside), a phenylpropanoid glycoside isolated from Rhodiola rosea, has been reported to have a broad spectrum of pharmacological properties. Salidroside is a prolyl endopeptidase inhibitor. Salidroside alleviates cachexia symptoms in mouse models of cancer cachexia via activating mTOR signalling. Salidroside protects dopaminergic neurons by enhancing PINK1/Parkin-mediated mitophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-568-8