Learn More
Growcells SALINE 9G/L 1L EA
Supplier:  Growcells MSDW1000
Encompass_Preferred
0.9% Sodium chloride aqueous solution, 18.2 megOhm water, 0.2 micron sterile filtered and steam sterilized in final packaging.
Catalog No. 50-103-4817