Learn More
Selleck Chemical LLC Salinomycin
SDP
Supplier:  Selleck Chemical LLC S81291G
Encompass_Preferred
Salinomycin traditionally used as an anti-coccidial drug has recently been shown to possess anti-cancer and anti-cancer stem cell (CSC) effects
Catalog No. 50-000-00136