Learn More
Medchemexpress LLC Salinomycin | 53003-10-4 | MFCD25541652 | 99.7% | 750.9986 g/mol | C42H70O11 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1559710MG
Encompass_Preferred
Salinomycin is a polyether potassium ionophore antibiotic used as a research reagent. It inhibits gram-positive bacterial growth and has been reported to block Wnt/β-catenin signaling and to show selective activity against cancer stem cells. Supplied as a solid powder for laboratory use; handle according to the safety data sheet.
  • High chemical purity suitable for research applications.
  • Powder form for easy dissolution and storage.
  • Potassium ionophore activity for ion transport studies.
  • Inhibits Wnt/β-catenin signaling for cell biology research.
  • Suitable for in vitro studies of cancer stem cells.
  • Supplied with a safety data sheet for safe handling.
Catalog No. 50-002-87747