Learn More
Medchemexpress LLC Salmeterol-d3 | 497063-94-2 | 98.0% | C25H34D3NO4 | 10MG

Supplier: Medchemexpress LLC HY13511910MG
Salmeterol-d3 is the deuterium-labeled analogue of salmeterol used as an analytical reference standard and for pharmacological research. It is supplied as a white to off-white solid with documented solubility and storage recommendations suitable for in vitro assays and in vivo formulations.
- Deuterium-labeled reference standard.
- Purity 98.0%.
- Molecular formula C25H34D3NO4.
- High solubility in DMSO (100 mg/mL).
- Stable as powder at -20°C for long-term storage.
- Suitable as internal standard for quantitative mass spectrometry.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.