Learn More
Selleck Chemical LLC Salmeterol Xinafoate S4296-50mg
SDP
Supplier:  Selleck Chemical LLC S429650MG
Encompass_Preferred
Salmeterol Xinafoate(GR 33343X xinafoate Salmetedur) is a long-acting 2-adrenergic receptor agonist with anti-inflammatory effects used in the treatment of asthma symptoms and chronic obstructive pulmonary disease (COPD) symptoms
Catalog No. 50-310-9429