Learn More
Medchemexpress LLC Salmeterol xinafoate | 94749-08-3 | 99.9% | 603.75 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1745310MM/1ML
Encompass_Preferred
Salmeterol xinafoate is a potent and selective human β2 adrenoceptor agonist. It shows potent stimulation of cAMP accumulation in CHO cells expressing human β2, β1 and β3 adrenoceptors.
  • Potent and selective human β2 adrenoceptor agonist.
  • Inhibits human T lymphocyte proliferation.
  • Therapeutic effects in mice with chronic obstructive pulmonary disease (COPD).
Catalog No. 50-003-86680