Learn More
Agilent Technologies Sonicated Salmon Sperm DNA, 10 x 1 mL
Supplier:  Agilent Technologies 201190
Encompass
A critical blocking agent when performing hybridization experiments Minimizes nonspecific interactions in hybridization experiments Molecular weight ranges from 300 bp to 2000 bp PURITY: DNase free CONCENTRATION: 10 mg/ml STORAGE CONDITIONS: -20°C
Catalog No. 50-125-193