Learn More
ATCC ATCC Salmonella enterica Panel (MP-15) - 5 Strains, Freeze-dried, 5 Vials
Supplier:  ATCC MP15
Encompass
The ATCC Salmonella enterica Panel (MP-15) is a curated set of five authenticated Salmonella enterica strains representing serovars commonly associated with contaminated food and water. Supplied freeze-dried in separate vials, the panel is intended for laboratory research applications including method development, assay qualification, and reference testing.
  • Five authenticated Salmonella enterica strains representing common serovars: Choleraesuis, Enteritidis, Newport, Typhi, Typhimurium.
  • Freeze-dried format for stable storage and transport.
  • Supplied as five individual vials for separate use.
  • Storage conditions: 2°C to 8°C.
  • Intended for laboratory research use, method development, and assay qualification.
Catalog No. NC3036795