Learn More
ATCC Salmonella enterica; subsp. enterica; serovar Enteritidis
Supplier:  ATCC 4931
Encompass
Salmonella enterica subsp. enterica serovar Enteritidis strain NCTC 4444 was isolated from a case of gastroenteritis in Copenhagen. This whole-genome sequenced bacterial strain has applications in emerging infectious disease research and enteric disease research.
Catalog No. 50-238-2761
 Restricted Item