Learn More
ATCC Salmonella enterica; subsp. enterica serovar; Typhimurium GFP
Supplier:  ATCC 14028GFP
Encompass
This clone was derived from ATCC 14028 and contains a multicopy vector encoding the green fluorescent protein (Aequorea victoria) GFPmut3. This gene is expressed under the control of the Plac promoter (Escherichia coli). This construct has been designed for Gram-negative bacteria fluorescence labeling. Ampicillin resistance gene (bla) encoded on a plasmid. Confers resistance to 100 μg/mL ampicillin.
Catalog No. 50-238-3779